Learn More
Sino Biological Monkeypox Virus (MPXV) L1R Protein (His Tag)
Supplier:  Sino Biological 40889V07E100
Encompass_Preferred
A DNA sequence encoding the Monkeypox Virus (MPXV) L1R Protein (EPI_ISL_13052264) (Met1-Lys152) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-6522