Learn More
Sino Biological Monkeypox Virus (MPXV) Protein A35 (His Tag)
Supplier:  Sino Biological 40886V07E100
Encompass_Preferred
A DNA sequence encoding the Monkeypox Virus (MPXV) Protein A35 (EPI_ISL_13052264) (Arg58-Thr181) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-9315