Learn More
Sino Biological Monkeypox Virus (MPXV) Protein A29 (His Tag)
Supplier:  Sino Biological 40891V08E100
Encompass_Preferred
A DNA sequence encoding the Monkeypox Virus (MPXV) Protein A29 (QNP13733.1) (Ser21-Glu110) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6523