Learn More
ATCC Monkeypox virus; Isolate: hMPXV/USA/MA001/2022
Catalog No. 502383613
Encompass
Monkeypox virus strain hMPXV/USA/MA001/2022 is propagated in BSC40 cells (ATCC CRL-2761). This virus was isolated in Massachusetts from a human in May 2022 and it belongs to Clade IIb and lineage B.1.This product requires a VS 16-6A permit, which can be downloaded at the USDA website here. Please submit the completed permit form to SalesPermits@atcc.org with your ATCC sales order number and/or your purchase number noted in the subject line and body of the email.
Catalog No. 50-238-3613 Supplier ATCC Supplier No. VR3371
Monkeypox virus strain hMPXV/USA/MA001/2022 is propagated in BSC40 cells (ATCC CRL-2761). This virus was isolated in Massachusetts from a human in May 2022 and it belongs to Clade IIb and lineage B.1.This product requires a VS 16-6A permit, which can be downloaded at the USDA website here. Please submit the completed permit form to SalesPermits@atcc.org with your ATCC sales order number and/or your purchase number noted in the subject line and body of the email.