Learn More
Medchemexpress LLC Mono-2-ethyl-5-carboxypentyl terephthalate-d4 | 1684398-42-2 | >85.0% | 312.35 g·mol⁻¹ | C16H16D4O6 | 1 MG
SDP
Catalog No. 5000284928
Encompass_Preferred
Mono-2-ethyl-5-carboxypentyl terephthalate-d4 is a deuterium-labeled analogue of mono-2-ethyl-5-carboxypentyl terephthalate supplied for use as an analytical tracer and internal standard. It enables accurate quantitation in trace analysis workflows and is intended for use in NMR, GC-MS, and LC-MS methods.
  • Isotope-labeled internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Deuterium labels provide clear mass shift for accurate detection.
  • Supplied as a small-quantity vial suitable for calibration and method development.
  • Certificate of Analysis and handling documentation available on request.
Catalog No. 50-002-84928 Supplier Medchemexpress LLC Supplier No. HYW587771S1MG
Mono-2-ethyl-5-carboxypentyl terephthalate-d4 is a deuterium-labeled analogue of mono-2-ethyl-5-carboxypentyl terephthalate supplied for use as an analytical tracer and internal standard. It enables accurate quantitation in trace analysis workflows and is intended for use in NMR, GC-MS, and LC-MS methods.
  • Isotope-labeled internal standard for quantitative analysis.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Deuterium labels provide clear mass shift for accurate detection.
  • Supplied as a small-quantity vial suitable for calibration and method development.
  • Certificate of Analysis and handling documentation available on request.