Learn More
Medchemexpress LLC Mono(2-ethylhexyl) phthalate | 4376-20-9 | MFCD00041500 | 97.0% | 278.34 g/mol | C16H22O4 | 1 ML
SDP
Catalog No. 5000301335
Encompass_Preferred
Mono(2-ethylhexyl) phthalate is an organic ester and the primary bioactive metabolite of di(2-ethylhexyl) phthalate (DEHP). It is commonly used as a reference standard and research reagent in toxicology, metabolism, and analytical chemistry. The compound has formula C16H22O4 and molecular weight 278.34 g/mol.
  • Primary bioactive metabolite of DEHP.
  • Commonly used as an analytical reference standard.
  • Reported purity 97%.
  • Molecular formula C16H22O4 and molecular weight 278.34 g/mol.
  • Used in toxicology, metabolism, and analytical studies.
Catalog No. 50-003-01335 Supplier Medchemexpress LLC Supplier No. HYW01839210MM/1ML
Mono(2-ethylhexyl) phthalate is an organic ester and the primary bioactive metabolite of di(2-ethylhexyl) phthalate (DEHP). It is commonly used as a reference standard and research reagent in toxicology, metabolism, and analytical chemistry. The compound has formula C16H22O4 and molecular weight 278.34 g/mol.
  • Primary bioactive metabolite of DEHP.
  • Commonly used as an analytical reference standard.
  • Reported purity 97%.
  • Molecular formula C16H22O4 and molecular weight 278.34 g/mol.
  • Used in toxicology, metabolism, and analytical studies.