Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich mono-2-(Methacryloyloxy)ethyl maleate

Catalog No. 483729100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-372-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-372-9100ML Supplier Sigma Aldrich Supplier No. 483729100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

mono-2-(Methacryloyloxy)ethyl maleate

Specifications

CAS 51978-15-5
Density 1.217 g/mL (at 25°C)
Molecular Formula C10H12O6
Linear Formula H2C=C(CH3)CO2CH2CH2O2CCH=CHCO2H
Refractive Index n20/D 1.483 (literature)
Quantity 100 mL
MDL Number MFCD00080563
Synonym (Z)-4-(2-(Methacryloyloxy)ethoxy)-4-oxobut-2-enoic acid; 2-Butenedioic Acid (Z)-Monophenyl Ester; Ethylene glycol methacrylate maleate; Mono-2-(methacryloyloxy)ethyl maleate
Molecular Weight (g/mol) 228.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.