Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich mono-2-(Methacryloyloxy)ethyl succinate

Catalog No. 454974250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-497-4250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-497-4250ML Supplier Sigma Aldrich Supplier No. 454974250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

mono-2-(Methacryloyloxy)ethyl succinate

Specifications

CAS 20882-04-6
Density 1.19 g/mL (at 25°C)
Boiling Point 267°C
Molecular Formula C10H14O6
Refractive Index n20/D 1.464 (literature)
Linear Formula H2C=C(CH3)CO2CH2CH2O2CCH2CH2CO2H
Quantity 250 mL
MDL Number MFCD00274302
Synonym 2-(Methacryloyloxy)ethyl hydrogen succinate; 2-Methacryloyloxyethyl succinate; 3-[2-(Methacryloyloxy)ethoxycarbonyl]propionic acid; 4-[2-(Methacryloyloxy)ethoxy]-4-oxobutanoic acid; Methacryloyloxyethyl succinate; Monomethacryloyloxyethyl succinate
Molecular Weight (g/mol) 230.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.