Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mono-(6-p-toluenesulfonyl)-β-cyclodextrin | 67217-55-4 | MFCD05864975 | 95.0% | 1289.17 g/mol | C49H76O37S | 500 MG
SDP

Catalog No. 5000282759
Encompass_Preferred

Mono-(6-p-toluenesulfonyl)-β-cyclodextrin is a tosylated derivative of β-cyclodextrin supplied as a white to off-white solid. It is used as a biochemical reagent and intermediate for selective functionalization at the 6-position of β-cyclodextrin, and is commonly applied in host-guest chemistry, derivatization, and synthetic modification in life-science research.

  • Purity 95.0%.
  • Appearance white to off-white solid.
  • Molecular formula C49H76O37S; molecular weight 1289.17 g/mol.
  • Enables selective functionalization at the 6-position of β-cyclodextrin.
  • Suitable for host-guest chemistry and derivatization studies.
  • Available in small lab pack sizes for research use.

Catalog No. 50-002-82759 Supplier Medchemexpress LLC Supplier No. HYW129395500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mono-(6-p-toluenesulfonyl)-β-cyclodextrin is a tosylated derivative of β-cyclodextrin supplied as a white to off-white solid. It is used as a biochemical reagent and intermediate for selective functionalization at the 6-position of β-cyclodextrin, and is commonly applied in host-guest chemistry, derivatization, and synthetic modification in life-science research.

  • Purity 95.0%.
  • Appearance white to off-white solid.
  • Molecular formula C49H76O37S; molecular weight 1289.17 g/mol.
  • Enables selective functionalization at the 6-position of β-cyclodextrin.
  • Suitable for host-guest chemistry and derivatization studies.
  • Available in small lab pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.