Learn More
Medchemexpress LLC Mono-(7-carboxy-4-methylheptyl) phthalate-d4 | 1332966-02-5 | >95.0% | 326.38 | C17H18D4O6 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYW588299S500UG
Encompass_Preferred
Mono-(7-carboxy-4-methylheptyl) phthalate-d4 is a deuterium-labeled reference standard of Mono-(7-carboxy-4-methylheptyl) phthalate used primarily as an internal standard in mass spectrometry and other analytical assays. It has chemical formula C17H18D4O6, molecular weight 326.38, and CAS number 1332966-02-5. The material is supplied in small, pre-weighed amounts (for example, 500 μg or 1 mg) and is intended for research use only.
  • Deuterium-labeled analog for reliable quantitation in MS assays.
  • Pre-weighed small-quantity vials suitable for analytical workflows.
  • Appropriate as an internal standard for phthalate metabolite analysis.
  • Purity reported greater than 95% for certified reference material variants.
  • Intended for research use only.
Catalog No. 50-002-84030