Learn More
Diagnostic Biosystems MONO AEC/PLUS Mono AEC/Plus
Catalog No. 502407627
Encompass_Preferred
Mono AEC/Plus is a single, highly stable, working AEC chromogen/substrate liquid solution. When used in conjunction with immunoperoxidase staining, AEC produces a red colored end product at the positive sites, yielding strong contrast when combined with a blue hematoxylin counter stain. AEC has been well accepted amongst pathologists because of its relatively less toxic nature as compared to DAB. Specimens stained using Mono AEC/Plus cannot be dehydrated in ethanol and hence need to be mounted in an aqueous-based mounting medium such as CC Mount (DBS catalog # K002).
Catalog No. 50-240-7627 Supplier Diagnostic Biosystems Supplier No. K050110
Mono AEC/Plus is a single, highly stable, working AEC chromogen/substrate liquid solution. When used in conjunction with immunoperoxidase staining, AEC produces a red colored end product at the positive sites, yielding strong contrast when combined with a blue hematoxylin counter stain. AEC has been well accepted amongst pathologists because of its relatively less toxic nature as compared to DAB. Specimens stained using Mono AEC/Plus cannot be dehydrated in ethanol and hence need to be mounted in an aqueous-based mounting medium such as CC Mount (DBS catalog # K002).