Learn More
Medchemexpress LLC HY-135297 100mg Medchemexpress, Mono-and diglycerides CAS: Purity:>98%
SDP
Catalog No. 501969871
Encompass_Preferred
Medchemexpress, HY-135297 100mg Mono-and diglycerides CAS: Mono-and diglycerides is formed by triglycerides being broken down by pancreatic lipase in the gastrointestinal lumen. Mono-and diglycerides is a food additive used as a nonionic emulsifier and mainly present in food fats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9871 Supplier Medchemexpress LLC Supplier No. HY135297100MG
Medchemexpress, HY-135297 100mg Mono-and diglycerides CAS: Mono-and diglycerides is formed by triglycerides being broken down by pancreatic lipase in the gastrointestinal lumen. Mono-and diglycerides is a food additive used as a nonionic emulsifier and mainly present in food fats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.