Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal Anti-Acetylated Tubulin antibody produced in mouse, MFCD00164512, 200 uL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T7451200UL
Encompass
Clone 6-11B-1, purified from hybridoma cell culture, Synonym: Acetylated Tubulin Antibody - Monoclonal Anti-Acetylated Tubulin antibody produced in mouse, Acetylated Tubulin Antibody. The antibody has been used to detect acetylated α-tubulins from many organisms including protista, plants, invertebrates, and vertebrates.
Catalog No. NC1247720