Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal ANTI-FLAG M2 antibody produced in mouse, 200 ug
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F1804200UG
Encompass
For highly sensitive and specific detection of FLAG fusion proteins by immunoblotting, immunoprecipitation (IP), immunohistochemisty, immunofluorescence and immunocyotchemistry. Optimized for single banded detection of FLAG fusion proteins in mammalian, plant, and bacterial expression systems.
Catalog No. NC1241958