Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mono-Butyl phthalate, MilliporeSigma™ Supelco™

Catalog No. 111012935 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2935 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2935 Supplier MilliporeSigma Supelco Supplier No. 30751100MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 131-70-4
Physical Form Neat
Linear Formula C12H14O4
MDL Number MFCD00043498
Synonym MBP; Phthalic acid butyl ester; mono-n-Butyl phthalate
Type Analytical Standard
Quantity 100 mg
Formula Weight 222.24
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.