Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mono-cyclohexyl phthalate-3,4,5,6-d4 | 1398066-18-6 | 99.1% | 252.30 g·mol-1 | C14H12D4O4 | 10 MG
SDP

Catalog No. 5000287675
Encompass_Preferred

Mono-cyclohexyl phthalate-3,4,5,6-d4 is a deuterium-labeled analogue of mono-cyclohexyl phthalate intended for use as a tracer and internal standard in quantitative analytical workflows. It is supplied as a solid powder with high isotopic purity and is suitable for NMR, GC-MS, and LC-MS applications.

  • High isotopic purity (99.1%) for accurate quantitation.
  • Molecular weight 252.30 g·mol-1 and formula C14H12D4O4.
  • Supplied as a solid powder with recommended cold storage for stability.
  • Recommended as a tracer and internal standard for NMR, GC-MS, and LC-MS analyses.
  • Available in small analytical packaging (10 mg) for laboratory use.

Catalog No. 50-002-87675 Supplier Medchemexpress LLC Supplier No. HY151911S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mono-cyclohexyl phthalate-3,4,5,6-d4 is a deuterium-labeled analogue of mono-cyclohexyl phthalate intended for use as a tracer and internal standard in quantitative analytical workflows. It is supplied as a solid powder with high isotopic purity and is suitable for NMR, GC-MS, and LC-MS applications.

  • High isotopic purity (99.1%) for accurate quantitation.
  • Molecular weight 252.30 g·mol-1 and formula C14H12D4O4.
  • Supplied as a solid powder with recommended cold storage for stability.
  • Recommended as a tracer and internal standard for NMR, GC-MS, and LC-MS analyses.
  • Available in small analytical packaging (10 mg) for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.