Learn More
Medchemexpress LLC Mono-Cyclohexyl Phthalate-3,4,5,6-d4 | 1398066-18-6 | C14H12D4O4 | 5 MG
SDP
Catalog No. 5000374149
Encompass_Preferred
Mono-Cyclohexyl Phthalate-3,4,5,6-d4 is the deuterium labeled Mono-Cyclohexyl Phthalate. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules as tracers
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-74149 Supplier Medchemexpress LLC Supplier No. HY151911S5MG
Mono-Cyclohexyl Phthalate-3,4,5,6-d4 is the deuterium labeled Mono-Cyclohexyl Phthalate. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules as tracers
  • Potential to affect pharmacokinetic and metabolic profiles of drugs