Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creative Bioarray MONO-MAC-1 cell line, Catalog No. CSC-C0353, Human, Acute monocytic leukemia (AML FAB M5), Suspension culture, Doubling time 40 hours, 1 x 10^6 cells/ml.

Catalog No. NC0660653
Encompass

The MONO-MAC-1 cell line is a human cell line sourced from the peripheral blood of a 64-year-old male diagnosed with acute monocytic leukemia (AML FAB M5). Known for its role in HIV-1 research and hematological studies, this less differentiated cell line supports productive HIV-1 infection, making it a critical model for examining monocytic functions and disease mechanisms.

  • Catalog number: CSC-C0353
  • Species: Human
  • Source: Peripheral blood from a patient with acute monocytic leukemia
  • Morphology: Single, round/multiformed cells in suspension
  • Doubling time: 30-50 hours
  • Mycoplasma status: Negative
  • Biosafety level: 1
  • Supports productive infection with HIV-1
  • Storage conditions: Frozen with medium and DMSO

Catalog No. NC0660653 Supplier Creative Bioarray Supplier No. CSCC0353
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MONO-MAC-1 cell line is a human cell line sourced from the peripheral blood of a 64-year-old male diagnosed with acute monocytic leukemia (AML FAB M5). Known for its role in HIV-1 research and hematological studies, this less differentiated cell line supports productive HIV-1 infection, making it a critical model for examining monocytic functions and disease mechanisms.

  • Catalog number: CSC-C0353
  • Species: Human
  • Source: Peripheral blood from a patient with acute monocytic leukemia
  • Morphology: Single, round/multiformed cells in suspension
  • Doubling time: 30-50 hours
  • Mycoplasma status: Negative
  • Biosafety level: 1
  • Supports productive infection with HIV-1
  • Storage conditions: Frozen with medium and DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.