Learn More
Cell Signaling Technology Mono-Methyl-Histone H3 (Lys9) Matched Antibody Pair, Unconjugated, 1 Kit
SDP
Catalog No. 503133826
Encompass
The Mono-Methyl-Histone H3 (Lys9) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, supporting various labeling methods including fluorophores, lanthanides, biotin, and beads.
  • Ideal for high-throughput platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8)
  • Provided BSA and azide free to facilitate custom conjugation
  • Validated for reactivity in human, mouse, rat, and monkey species
  • Includes 100 μg of capture antibody and 100 μg of detection antibody
Catalog No. 50-313-3826 Supplier Cell Signaling Technology Supplier No. 68690S
The Mono-Methyl-Histone H3 (Lys9) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, supporting various labeling methods including fluorophores, lanthanides, biotin, and beads.
  • Ideal for high-throughput platforms such as MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8)
  • Provided BSA and azide free to facilitate custom conjugation
  • Validated for reactivity in human, mouse, rat, and monkey species
  • Includes 100 μg of capture antibody and 100 μg of detection antibody