Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich mono-Methyl hydrogen succinate

Catalog No. M81101100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M81101100G 100 g
M8110125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M81101100G Supplier Sigma Aldrich Supplier No. M81101100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

mono-Methyl hydrogen succinate

Specifications

CAS 3878-55-5
Melting Point 54°C to 57°C (lit.)
Boiling Point 151°C (20 mmHg)
Molecular Formula C5H8O4
Linear Formula CH3OCOCH2CH2COOH
MDL Number MFCD00002788
Quantity 100 g
Synonym Monomethyl succinate; Succinic acid monomethyl ester
Molecular Weight (g/mol) 132.11
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.