Learn More
Accela Chembio Inc MF: C9H8O4 | MW: 180.16 | ?98% | Flash Point: 136? | BP: 331.6? at 760 mmHg.
Catalog No. 50354915
Encompass_Preferred
MF: C9H8O4 | MW: 180.16 | ?98% | Flash Point: 136? | BP: 331.6? at 760 mmHg.
Catalog No. 50-354-915 Supplier Accela Chembio Inc Supplier No. SY010493100G
MF: C9H8O4 | MW: 180.16 | ?98% | Flash Point: 136? | BP: 331.6? at 760 mmHg.