Learn More
Cole-Parmer Mono-Mold Standard Stir Bar with Spinning Ring, 3"L x 1/2" dia
Catalog No. NC1792897 Shop All Cole-Parmer Products
Encompass_Preferred
Mono-Mold magnetic stir bars are the standard all-purpose stir bar. Available with or without spinning rings; rings reduce contact area between stir bar and vessel, reducing friction and noise.
Catalog No. NC1792897 Supplier Cole-Parmer Supplier No. 0461230
Mono-Mold magnetic stir bars are the standard all-purpose stir bar. Available with or without spinning rings; rings reduce contact area between stir bar and vessel, reducing friction and noise.