Learn More
Cole-Parmer Mono-Mold Standard Stir Bar, 1-1/2"L x 3/8" dia
Supplier:  Cole-Parmer 0477050
Encompass_Preferred
Mono-Mold magnetic stir bars are the standard all-purpose stir bar. Available with or without spinning rings; rings reduce contact area between stir bar and vessel, reducing friction and noise.
Catalog No. NC1803671