Learn More
Medchemexpress LLC Mono-n-nonyl phthalate-3,4,5,6-d4 | 2514564-42-0 | ≥98.0% | 296.39 g·mol⁻¹ | C17H20D4O4 | 10 MG
SDP
Catalog No. 5000286201
Encompass_Preferred
Mono-n-Nonyl Phthalate-3,4,5,6-d4 is the deuterium-labeled (d4) analogue of mono-n-nonyl phthalate, provided as an analytical reference standard for mass spectrometry, isotopic tracing, and method development. It is intended for use as an internal standard in quantitative LC-MS/MS assays and environmental or biological phthalate analyses.
Catalog No. 50-002-86201 Supplier Medchemexpress LLC Supplier No. HY150863S10MG
Mono-n-Nonyl Phthalate-3,4,5,6-d4 is the deuterium-labeled (d4) analogue of mono-n-nonyl phthalate, provided as an analytical reference standard for mass spectrometry, isotopic tracing, and method development. It is intended for use as an internal standard in quantitative LC-MS/MS assays and environmental or biological phthalate analyses.