Learn More
Medchemexpress LLC Mono-n-octyl phthalate-d4 | 1398065-74-1 | MFCD22565813 | >95.0% | 282.37 g/mol | C16H18D4O4 | 5 MG
SDP
Catalog No. 5000262699
Encompass_Preferred
Monooctyl phthalate-d4 is a deuterium-labeled reference standard used as an internal standard and tracer in analytical chemistry workflows, including LC-MS, GC-MS, and NMR. Supplied as a solid, it supports method development and accurate quantitation in research applications. CAS 1398065-74-1; formula C16H18D4O4; molecular weight 282.37 g/mol.
Catalog No. 50-002-62699 Supplier Medchemexpress LLC Supplier No. HY133671S5MG
Monooctyl phthalate-d4 is a deuterium-labeled reference standard used as an internal standard and tracer in analytical chemistry workflows, including LC-MS, GC-MS, and NMR. Supplied as a solid, it supports method development and accurate quantitation in research applications. CAS 1398065-74-1; formula C16H18D4O4; molecular weight 282.37 g/mol.