Learn More
Cayman Chemical Monoacylglycerol Lipase Block
SDP
Catalog No. 502989717
Encompass_Preferred
Peptide Sequence: human monoacylglycerol lipase blocking peptide amino acids 1-14 (MPEESSPRRTPQSI) · To be used in conjunction with Cayman’s Monoacylglycerol Lipase Polyclonal Antibody (Item No. 100035) to block protein-antibody complex formation during analysis for monoacylglycerol lipase.
Catalog No. 50-298-9717 Supplier Cayman Chemical Supplier No. 3000141 EA
Peptide Sequence: human monoacylglycerol lipase blocking peptide amino acids 1-14 (MPEESSPRRTPQSI) · To be used in conjunction with Cayman’s Monoacylglycerol Lipase Polyclonal Antibody (Item No. 100035) to block protein-antibody complex formation during analysis for monoacylglycerol lipase.