Learn More
Cayman Chemical Monoacylglycerol Lipase (FL) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502886632
Encompass_Preferred
Immunogen: Purified recombinant human MAGL • Host: Rabbit • Species Reactivity: (+) Human and rat; other species not tested • Applications: ICC, IHC, and WB
Catalog No. 50-288-6632 Supplier Cayman Chemical Supplier No. 102121 EA
Immunogen: Purified recombinant human MAGL • Host: Rabbit • Species Reactivity: (+) Human and rat; other species not tested • Applications: ICC, IHC, and WB