Learn More
Cayman Chemical Monoacylglycerol Lipase Inhib
SDP
Catalog No. 502967209
Encompass_Preferred
Selectively and reversibly blocks MAGL activity (IC50s = 0.18 and 59 µM for MAGL and FAAH in mouse brain, respectively); does not inhibit ABHD6, ABHD12, CB1, or CB2 receptors (Kis > 10 µM); used to ameliorate disease progression in a mouse model of multiple sclerosis
Catalog No. 50-296-7209 Supplier Cayman Chemical Supplier No. 169645 MG
Selectively and reversibly blocks MAGL activity (IC50s = 0.18 and 59 µM for MAGL and FAAH in mouse brain, respectively); does not inhibit ABHD6, ABHD12, CB1, or CB2 receptors (Kis > 10 µM); used to ameliorate disease progression in a mouse model of multiple sclerosis