Learn More
Cayman Chemical Monoacylglycerol Lipase Polyc
SDP
Catalog No. 502886555
Encompass_Preferred
Immunogen: Synthetic peptide from human MAGL • Species Reactivity: (+) Human, bovine, mouse, and rat • Host: Rabbit • Applications: IHC (formalin-fixed paraffin-embedded tissue) and WB
Catalog No. 50-288-6555 Supplier Cayman Chemical Supplier No. 1000351 EA
Immunogen: Synthetic peptide from human MAGL • Species Reactivity: (+) Human, bovine, mouse, and rat • Host: Rabbit • Applications: IHC (formalin-fixed paraffin-embedded tissue) and WB