Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoammonium Glycyrrhizinate Hydrate 75.0+%, TCI America™
SDP

Catalog No. G01511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01511G Supplier TCI America Supplier No. G01511G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53956-04-0
Molecular Formula C42H65NO16
Molecular Weight (g/mol) 839.97
MDL Number MFCD00167400,MFCD00167400
InChI Key ILRKKHJEINIICQ-OOFFSTKBSA-N
Synonym Glycyrrhizic Acid Monoammonium Salt, Glycyrrhizin Monoammonium Salt
PubChem CID 131676200
IUPAC Name (2S,3S,4S,5R,6S)-6-{[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-5-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3,4-dihydroxyoxane-2-carboxylic acid amine
SMILES N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 1 g
Formula Weight 839.97
Percent Purity ≥75.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Monoammonium Glycyrrhizinate Hydrate
EINECSNumber (9)-0398
RTECSNumber LZ6500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.