Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobasic Sodium Phosphate, Dihydrate, USP, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18604665 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
1 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-665 50 kg Poly Bottle
18-604-662 500 g Poly Bottle
18-604-663 1 kg Poly Bottle
18-604-664 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-665 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S193050KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

S1930, 13472-35-0, NaH2PO4·2H2O

Spectrum™ Chemical Monobasic Sodium Phosphate, Dihydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
Molecular Weight (g/mol) 156.01
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
IUPAC Name sodium dihydrate dihydrogen phosphate
SMILES O.O.[Na+].OP(O)([O-])=O

Specifications

pH 4.1 to 4.5
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 98 to 103%
Grade USP
Packaging Poly Bottle
Chemical Name or Material Monobasic Sodium Phosphate, Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.