Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604667 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-667 2.5 kg Poly Bottle
18-604-668 12 kg Poly Pail
18-604-670 25 kg Fiber Drum
18-604-669 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-667 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO1302.5KG
Add to Cart
Add to Cart

SO130, 10049-21-5, NaH2PO4·H2O

Spectrum™ Chemical Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP - All Spectrum Chemical USP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

pH 4.2 to 4.5
CAS Max % 1
Quantity 2.5 kg
Percent Purity 98 to 100.5%
Grade Multi-Compendial
Loss on Drying 11.5-14.5%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.