Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604670 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-670 25 kg Poly Bottle
18-604-667 2.5 kg Poly Bottle
18-604-668 12 kg Poly Bottle
18-604-669 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-670 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO13025KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SO130, 10049-21-5, NaH2PO4·H2O

Spectrum™ Chemical Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP - All Spectrum Chemical USP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

pH 4.2 to 4.5
CAS Max % 1
Physical Form Granular/Lump
Quantity 25 kg
Percent Purity 98 to 100.5%
Grade Multi-Compendial
Loss on Drying 11.5-14.5%
Packaging Poly Bottle
Chemical Name or Material Monobasic Sodium Phosphate, Monohydrate, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.