Learn More
Apexbio Technology LLC Monobenzone 103-16-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000095752
Encompass_Preferred
Monobenzone is a small-molecule inhibitor targeting tyrosinase It is designed to inhibit the enzyme tyrosinase thereby disrupting the melanin biosynthesis pathway and reducing melanin production Monobenzone exerts its biological activity primarily through inhibition of tyrosinase activity Based on these pharmacological properties Monobenzone holds research potential in studies on melanoma vitiligo mechanisms melanin regulation and in generating depigmented animal models and cellular assays for investigating pigment synthesis melanocyte biology and related autoimmune responses
Catalog No. 50-000-95752 Supplier Apexbio Technology LLC Supplier No. A84785.1
Monobenzone is a small-molecule inhibitor targeting tyrosinase It is designed to inhibit the enzyme tyrosinase thereby disrupting the melanin biosynthesis pathway and reducing melanin production Monobenzone exerts its biological activity primarily through inhibition of tyrosinase activity Based on these pharmacological properties Monobenzone holds research potential in studies on melanoma vitiligo mechanisms melanin regulation and in generating depigmented animal models and cellular assays for investigating pigment synthesis melanocyte biology and related autoimmune responses