Learn More
Medchemexpress LLC Monobenzone | 103-16-2 | 99.9% | 200.24 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3027210G
Encompass_Preferred
Monobenzone is a potent skin depigmenting agent that induces depigmentation and shows potential for vitiligo research. It is also a potent inhibitor of RNR (Ribonucleotide reductase) enzyme activity by targeting RRM2 (a regulatory small subunit M2 of RNR) protein, demonstrating significant anti-leukemia efficacy both in vitro and in vivo. It inhibits acute myeloid leukemia (AML) cells proliferation and DNA synthesis, and induces cell cycle arrest and apoptosis.
  • Potent skin depigmenting agent
  • Induces depigmentation
  • Exhibits good potential for vitiligo research
  • Potent inhibitor of RNR enzyme activity
  • Has significant anti-leukemia efficacy in vitro and in vivo
  • Inhibits AML cells proliferation and DNA synthesis
  • Induces cell cycle arrest and apoptosis
Catalog No. 50-004-01449