Learn More
Medchemexpress LLC Monobenzone | 103-16-2 | 99.9% | 200.24 | 50 G
SDP
Catalog No. 5000400432
Encompass_Preferred
Monobenzone is a potent skin depigmenting agent that induces depigmentation and shows potential for vitiligo research. It is also a potent inhibitor of RNR (Ribonucleotide reductase) enzyme activity by targeting RRM2 protein, demonstrating significant anti-leukemia efficacy in vitro and in vivo. It inhibits acute myeloid leukemia (AML) cell proliferation and DNA synthesis, and induces cell cycle arrest and apoptosis.
  • Potent skin depigmenting agent.
  • Induces depigmentation.
  • Shows potential for vitiligo research.
  • Potent inhibitor of RNR enzyme activity by targeting RRM2 protein.
  • Demonstrates significant anti-leukemia efficacy in vitro and in vivo.
  • Inhibits acute myeloid leukemia (AML) cell proliferation and DNA synthesis.
  • Induces cell cycle arrest and apoptosis.
Catalog No. 50-004-00432 Supplier Medchemexpress LLC Supplier No. HY3027250G
Add to Cart
Add to Cart
Monobenzone is a potent skin depigmenting agent that induces depigmentation and shows potential for vitiligo research. It is also a potent inhibitor of RNR (Ribonucleotide reductase) enzyme activity by targeting RRM2 protein, demonstrating significant anti-leukemia efficacy in vitro and in vivo. It inhibits acute myeloid leukemia (AML) cell proliferation and DNA synthesis, and induces cell cycle arrest and apoptosis.
  • Potent skin depigmenting agent.
  • Induces depigmentation.
  • Shows potential for vitiligo research.
  • Potent inhibitor of RNR enzyme activity by targeting RRM2 protein.
  • Demonstrates significant anti-leukemia efficacy in vitro and in vivo.
  • Inhibits acute myeloid leukemia (AML) cell proliferation and DNA synthesis.
  • Induces cell cycle arrest and apoptosis.