Learn More
Apexbio Technology LLC MONOBENZYL-PHTHALATE-1G
SDP
Catalog No. 5000568855
Encompass_Preferred
Monobenzyl phthalate (CAS No 2528-16-7) is investigated primarily in the context of toxicology and environmental health research It originates as a metabolite of phthalate esters commonly used in manufacturing plastics Its mechanism of action involves disrupting endocrine signaling pathways particularly affecting hormone receptors In vitro studies often demonstrate activity within the nanomolar to low micromolar range Typical applications include assessments in cell models such as hepatocytes or reproductive cells as well as in vivo animal studies to evaluate its impact on development and reproduction It is also utilized in drug screening for its potential endocrine-disrupting effects Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68855 Supplier Apexbio Technology LLC Supplier No. C65901000
Monobenzyl phthalate (CAS No 2528-16-7) is investigated primarily in the context of toxicology and environmental health research It originates as a metabolite of phthalate esters commonly used in manufacturing plastics Its mechanism of action involves disrupting endocrine signaling pathways particularly affecting hormone receptors In vitro studies often demonstrate activity within the nanomolar to low micromolar range Typical applications include assessments in cell models such as hepatocytes or reproductive cells as well as in vivo animal studies to evaluate its impact on development and reproduction It is also utilized in drug screening for its potential endocrine-disrupting effects Experimental concentrations typically depend on the specific research design