Learn More
Medchemexpress LLC Monobenzyl phthalate | 2528-16-7 | 99.9% | 256.25 | 10 MG
SDP
Catalog No. 5000453744
Encompass_Preferred
Monobenzyl phthalate (2-((Benzyloxy)carbonyl)benzoic acid) is a urinary metabolite found after exposure to phthalates such as diethylhexyl phthalate (DEHP). This product is designed strictly for research use.
  • Urinary metabolite of phthalate exposure
  • Research use only
  • Stable under recommended storage conditions
Catalog No. 50-004-53744 Supplier Medchemexpress LLC Supplier No. HYW011848S10MG
Monobenzyl phthalate (2-((Benzyloxy)carbonyl)benzoic acid) is a urinary metabolite found after exposure to phthalates such as diethylhexyl phthalate (DEHP). This product is designed strictly for research use.
  • Urinary metabolite of phthalate exposure
  • Research use only
  • Stable under recommended storage conditions