Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monobenzyl phthalate | 2528-16-7 | 99.9% | 256.26 | 500 MG
SDP

Catalog No. 5000455016
Encompass_Preferred

Monobenzyl phthalate (2-((Benzyloxy)carbonyl)benzoic acid) is a urinary metabolite found after exposure to phthalates, such as diethylhexyl phthalate (DEHP). It is for research use only. This product offers high purity and is supplied as a white to off-white solid with specific storage conditions to maintain stability.

  • High purity of 99.88%.
  • Molecular weight of 256.25.
  • Provided as a white to off-white solid.
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage for solution: -80°C for 6 months, -20°C for 1 month.
  • Soluble in DMSO.

Catalog No. 50-004-55016 Supplier Medchemexpress LLC Supplier No. HYW011848500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monobenzyl phthalate (2-((Benzyloxy)carbonyl)benzoic acid) is a urinary metabolite found after exposure to phthalates, such as diethylhexyl phthalate (DEHP). It is for research use only. This product offers high purity and is supplied as a white to off-white solid with specific storage conditions to maintain stability.

  • High purity of 99.88%.
  • Molecular weight of 256.25.
  • Provided as a white to off-white solid.
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage for solution: -80°C for 6 months, -20°C for 1 month.
  • Soluble in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.