Learn More
Medchemexpress LLC Monobenzyl terephthalate | 18520-63-3 | MFCD01464148 | 99.9% | 250mg
SDP
Catalog No. 5000267607
Encompass_Preferred
Monobenzyl Terephthalate (4-((Benzyloxy)carbonyl)benzoic acid) is a drug intermediate for synthesis of various active compounds
Catalog No. 50-002-67607 Supplier Medchemexpress LLC Supplier No. HYZ1081250MG
Monobenzyl Terephthalate (4-((Benzyloxy)carbonyl)benzoic acid) is a drug intermediate for synthesis of various active compounds