Learn More
Echelon Biosciences Research Labs MONOBROMABIMANE 25 mg
SDP
Supplier:  Echelon Biosciences Research Labs F003025MG
Encompass
Monobromobimane reacts with thiols, sulfide, thiosulfate, and sulfite to generate a fluorescent product with an excitation/ emission spectra of 394/ 490 nm. This item is non returnable
Catalog No. 50-720-325