Learn More
Enzo Life Sciences Monobromobimane (ultra pure) (25mg). CAS: 71418-44-5
SDP
Supplier:  Enzo Life Sciences ENZ52501
Encompass_Preferred
Monobromobimane (mBBr) is a thiol-reactive fluorescent probe used for the determination of the redox status of low molecular weight and protein thiols in biological systems. Monobromobimane-based in situ derivatization results in maximal recovery of both free, reduced low molecular weight and monobromobimane-accessible protein thiols. The quantitation of the corresponding adducts of protein thiols is achieved by fluorescence spectroscopy, following protein precipitation. The reagent is particularly useful for quantifying glutathione. Wavelength Maxima: Excitation 395nm, Emission 490nm Alternative name: mBBr, 3-(Bromomethyl)-2,5,6-trimethyl-1H,7H-pyrazolo(1,2-alpha) pyrazole-1,7-dione. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2642