Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monobromobimane (mBBr), FluoroPure™ grade

Catalog No. M20381
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20381 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20381 Supplier Invitrogen™ Supplier No. M20381
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Because it is manufactured at our ISO 13485—certified facilities in Eugene, Oregon, we can guarantee that FluoroPure grade monobromobimane is greater than or equal to 98% pure by HPLC.

Because it is manufactured at our ISO 13485-certified facilities in Eugene, Oregon, we can guarantee that FluoroPure grade monobromobimane is greater than or equal to 98% pure by HPLC.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type Monobromobimane
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Line FluoroPure

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.