Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monobutyl phthalate | 131-70-4 | 1 ML
SDP

Catalog No. 5000419878
Encompass_Preferred

Monobutyl phthalate is identified as a significant metabolite of dibutyl phthalate (DBP) and exhibits antiandrogenic properties. It is also classified as an embryotoxicant. This product is intended for research use only and is not sold to patients. The compound appears as a solid, ranging from white to off-white in color, with a molecular weight of 222.24 and a chemical formula of C12H14O4. Its structure is classified under ketones, aldehydes, and acids, and it is an endogenous metabolite. Monobutyl phthalate has a purity of 99.72%.

  • Possesses antiandrogenic properties as a major metabolite of dibutyl phthalate (DBP)
  • Identified as an embryotoxicant
  • Available with a purity of 99.72%
  • Primarily for research applications
  • Soluble in DMSO at 100 mg/mL (449.96 mM), requiring sonication
  • Storage: powder stable for 3 years at -20°C and 2 years at 4°C; in solvent stable for 6 months at -80°C and 1 month at -20°C

Catalog No. 50-004-19878 Supplier Medchemexpress LLC Supplier No. HYN714310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monobutyl phthalate is identified as a significant metabolite of dibutyl phthalate (DBP) and exhibits antiandrogenic properties. It is also classified as an embryotoxicant. This product is intended for research use only and is not sold to patients. The compound appears as a solid, ranging from white to off-white in color, with a molecular weight of 222.24 and a chemical formula of C12H14O4. Its structure is classified under ketones, aldehydes, and acids, and it is an endogenous metabolite. Monobutyl phthalate has a purity of 99.72%.

  • Possesses antiandrogenic properties as a major metabolite of dibutyl phthalate (DBP)
  • Identified as an embryotoxicant
  • Available with a purity of 99.72%
  • Primarily for research applications
  • Soluble in DMSO at 100 mg/mL (449.96 mM), requiring sonication
  • Storage: powder stable for 3 years at -20°C and 2 years at 4°C; in solvent stable for 6 months at -80°C and 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.