Learn More
Medchemexpress LLC Monobutyl phthalate | 131-70-4 | MFCD00043498 | 100 MG
SDP
Catalog No. 5000417971
Encompass_Preferred
Monobutyl phthalate, a major metabolite of dibutyl phthalate (DBP), exhibits antiandrogenic effects and is classified as an embryotoxicant.
  • Exhibits antiandrogenic effects.
  • It is an embryotoxicant.
  • For research use only.
  • Purity of 99.72%.
  • Available in solid and solution forms.
Catalog No. 50-004-17971 Supplier Medchemexpress LLC Supplier No. HYN7143R100MG
Monobutyl phthalate, a major metabolite of dibutyl phthalate (DBP), exhibits antiandrogenic effects and is classified as an embryotoxicant.
  • Exhibits antiandrogenic effects.
  • It is an embryotoxicant.
  • For research use only.
  • Purity of 99.72%.
  • Available in solid and solution forms.