Learn More
Medchemexpress LLC Monobutyl phthalate-d4 | 478954-81-3 | 10 MG
SDP
Catalog No. 5000417563
Encompass_Preferred
Monobutyl phthalate-d4 is a deuterium-labeled form of monobutyl phthalate. Monobutyl phthalate, a key metabolite of dibutyl phthalate (DBP), is recognized for its antiandrogenic effects and as an embryotoxicant. Deuteration, which involves incorporating stable heavy isotopes, is often used for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer.
  • Functions as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Features stable heavy isotopes for drug development quantitation.
Catalog No. 50-004-17563 Supplier Medchemexpress LLC Supplier No. HYN7143S10MG
Monobutyl phthalate-d4 is a deuterium-labeled form of monobutyl phthalate. Monobutyl phthalate, a key metabolite of dibutyl phthalate (DBP), is recognized for its antiandrogenic effects and as an embryotoxicant. Deuteration, which involves incorporating stable heavy isotopes, is often used for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer.
  • Functions as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Features stable heavy isotopes for drug development quantitation.