Learn More
Medchemexpress LLC Monobutyl phthalate (Standard) | 131-70-4 | 25 MG
SDP
Catalog No. 5000422002
Encompass_Preferred
Monobutyl phthalate (Standard) is the analytical standard of Monobutyl phthalate. Intended for research and analytical applications, it is a major metabolite of dibutyl phthalate (DBP), known for its antiandrogenic effects and as an embryotoxicant.
  • Analytical standard
  • Used for research and analytical applications
  • Major metabolite of dibutyl phthalate (DBP)
  • Possesses antiandrogenic effects
  • Functions as an embryotoxicant
  • Commonly used in HPLC, GC, and MS experiments
Catalog No. 50-004-22002 Supplier Medchemexpress LLC Supplier No. 25MGHYN7143R
Monobutyl phthalate (Standard) is the analytical standard of Monobutyl phthalate. Intended for research and analytical applications, it is a major metabolite of dibutyl phthalate (DBP), known for its antiandrogenic effects and as an embryotoxicant.
  • Analytical standard
  • Used for research and analytical applications
  • Major metabolite of dibutyl phthalate (DBP)
  • Possesses antiandrogenic effects
  • Functions as an embryotoxicant
  • Commonly used in HPLC, GC, and MS experiments