Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monobutyl phthalate (Standard) | 131-70-4 | 250 MG
SDP

Catalog No. 5000422018
Encompass_Preferred

Monobutyl phthalate (Standard) is the analytical standard of Monobutyl phthalate. This product is intended for research and analytical applications. Monobutyl phthalate, a major metabolite of dibutyl phthalate (DBP), possesses antiandrogenic effects and is an embryotoxicant.

  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Molecular weight: 222.24
  • Formula: C12H14O4
  • CAS No.: 131-70-4
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: O=C(O)C1=CC=CC=C1C(OCCCC)=O
  • Structure classification: Ketones, Aldehydes, Acids
  • Initial source: Endogenous metabolite
  • Purity: 99.86%

Catalog No. 50-004-22018 Supplier Medchemexpress LLC Supplier No. 250MGHYN7143R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monobutyl phthalate (Standard) is the analytical standard of Monobutyl phthalate. This product is intended for research and analytical applications. Monobutyl phthalate, a major metabolite of dibutyl phthalate (DBP), possesses antiandrogenic effects and is an embryotoxicant.

  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Molecular weight: 222.24
  • Formula: C12H14O4
  • CAS No.: 131-70-4
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: O=C(O)C1=CC=CC=C1C(OCCCC)=O
  • Structure classification: Ketones, Aldehydes, Acids
  • Initial source: Endogenous metabolite
  • Purity: 99.86%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.