Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobutyltin Oxide, TCI America™
SDP

Catalog No. M074425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M074425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M074425G Supplier TCI America Supplier No. M074425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2273-43-0
Molecular Formula C4H10O2Sn
Molecular Weight (g/mol) 208.83
MDL Number MFCD00013927
InChI Key WIHMDCQAEONXND-UHFFFAOYSA-M
Synonym stannane, butylhydroxyoxo, butyltin hydroxide oxide, butylstannoic acid, butylstannonic acid, 1-butanestannonic acid, n-butyltin hydroxide oxide, butylhydroxytin oxide, butylhydroxyoxostannane, unii-g34wda7z2e, butylenestannonic acid
PubChem CID 16767
IUPAC Name butyl(hydroxy)stannanone
SMILES CCCC[Sn](O)=O

Specifications

Color White
Quantity 25 g
UN Number 3146
Formula Weight 208.83
Physical Form Crystalline Powder
Chemical Name or Material Monobutyltin Oxide
EINECSNumber (2)-2269
RTECSNumber WH6770000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.