Learn More
Medchemexpress LLC Monocaprylin | 26402-26-6 | 98.0% | 218.29 | 100 MG
SDP
Catalog No. 5000347883
Encompass_Preferred
Monocaprylin (Glyceryl monocaprylate), a monoglyceride of caprylic acid, demonstrates excellent antibacterial activity. This compound inhibits various foodborne pathogenic and spoilage microorganisms, suggesting its potential as an alternative food preservative in research.
  • Exhibits excellent antibacterial activity.
  • Inhibits a variety of foodborne pathogenic and spoilage microorganisms.
  • Has potential for use as an alternative food preservative in research.
  • Reduces five specific pathogens (Strep. agalactiae, Strep. dysgalactiae, Strep. uberis, Staph. aureus, and E. coli) by over 5.0 log cfu/mL within six hours of incubation at 25 mM and 50 mM concentrations.
Catalog No. 50-003-47883 Supplier Medchemexpress LLC Supplier No. HY138650100MG
Monocaprylin (Glyceryl monocaprylate), a monoglyceride of caprylic acid, demonstrates excellent antibacterial activity. This compound inhibits various foodborne pathogenic and spoilage microorganisms, suggesting its potential as an alternative food preservative in research.
  • Exhibits excellent antibacterial activity.
  • Inhibits a variety of foodborne pathogenic and spoilage microorganisms.
  • Has potential for use as an alternative food preservative in research.
  • Reduces five specific pathogens (Strep. agalactiae, Strep. dysgalactiae, Strep. uberis, Staph. aureus, and E. coli) by over 5.0 log cfu/mL within six hours of incubation at 25 mM and 50 mM concentrations.